1,5-Diaminopentan
Lieferant: CUSTOM MADE CHEMICALS LAB
|
Gefahr
|
Synonyme:
Pentamethylendiamin, Cadavarin, 1,5-Pentandiamin
SB65012.250MG
CMPLSB65012.250MGEA
0
EUR
CMPLSB65012.250MG
1,5-Diaminopentan
1,5-Diaminopentan
Standards: ISO 17034, ISO 17025, ISO 9001.
Bestellinformation: Store in a refrigerator at temperatures between 2 to 8 °C.
Formel:
C₅H₁₄N₂ Molecular Weight: 102,18 g/mol Siedepunkt: 178,..180 °C Schmelzpunkt: 9 °C Dichte: 0,873 g/cm³ Flammpunkt: 62 °C |
MDL:
MFCD00008239 CAS-Nummer: 462-94-2 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...