Nitrogen (Total) Standard Solution, CRM
Lieferant: CUSTOM MADE CHEMICALS LAB
TN0100.L1
CMPLTN0100.L1EA
0
EUR
CMPLTN0100.L1
Nitrogen (Total) Standard Solution, CRM
Standards
Umweltanalyse-Standards
Total nitrogen standard (100 mg/L) - for photometric applications.
Standards: EN 25663 (ISO 5663) and ISO 7150. ISO 17034, ISO 17025, ISO 9001.
Bestellinformation: Shelf life on ship date: 24 Months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.