Methyltetrahydropyran-4-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl tetrahydro-2H-pyran-4-carboxylate

AM-1008-5G AM-1008-100G AM-1008-25G
COBBAM-1008-5GEA 0 EUR
COBBAM-1008-5G COBBAM-1008-100G COBBAM-1008-25G
Methyltetrahydropyran-4-carboxylat
Methyltetrahydropyran-4-carboxylat
Formel: C₇H₁₂O₃
MDL: MFCD01075170
CAS-Nummer: 110238-91-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR