1-Boc-4-piperidon

Lieferant: COMBI-BLOCKS

Synonyme: 4-Oxo-piperidine-1-carboxylic acidtert-butyl ester, N-BOC-4-Piperidone, N-tert-BOC-4-Piperidinone

AM-1027-25G AM-1027-5G AM-1027-100G
COBBAM-1027-25GEA 0 EUR
COBBAM-1027-25G COBBAM-1027-5G COBBAM-1027-100G
1-Boc-4-piperidon
1-Boc-4-piperidon
Formel: C₁₀H₁₇NO₃
MDL: MFCD00151800
CAS-Nummer: 79099-07-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR