1-Boc-piperidin-4-carbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid
AM-1030-5G
AM-1030-25G
AM-1030-100G
COBBAM-1030-5GEA
0
EUR
COBBAM-1030-5G
COBBAM-1030-25G
COBBAM-1030-100G
1-Boc-piperidin-4-carbonsäure
1-Boc-piperidin-4-carbonsäure
Formel:
C₁₁H₁₉NO₄ |
MDL:
MFCD00076999 CAS-Nummer: 84358-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...