(Tetrahydropyran-4-yliden)acetonitril

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Dihydro-2H-pyran-4(3H)-ylidene)acetonitrile

AM-1621-5G AM-1621-1G
COBBAM-1621-5GEA 0 EUR
COBBAM-1621-5G COBBAM-1621-1G
(Tetrahydropyran-4-yliden)acetonitril
(Tetrahydropyran-4-yliden)acetonitril
Formel: C₇H₉NO
MDL: MFCD09800849
CAS-Nummer: 204651-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR