4-Amino-1-Boc-piperidin

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 4-Amino-1-piperidinecarboxylate, 4-Amino-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-aminopiperidine-1-carboxylate, N-Boc-4-piperidineamine, tert-Butyl 4-amino-1-piperidinecarboxylate, 4-Amino-1-Boc-piperidine, 4-Amino-1-piperidinecarboxylic acid tert-Butyl Ester

AM-1626-5G AM-1626-25G AM-1626-1G AM-1626-100G
COBBAM-1626-5GEA 0 EUR
COBBAM-1626-5G COBBAM-1626-25G COBBAM-1626-1G COBBAM-1626-100G
4-Amino-1-Boc-piperidin
4-Amino-1-Boc-piperidin
Formel: C₁₀H₂₀N₂O₂
MDL: MFCD01076201
CAS-Nummer: 87120-72-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR