Methyltetrahydropyran-4-ylacetat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 2-(tetrahydro-2H-pyran-4-yl)acetate
AM-1629-1G
AM-1629-25G
AM-1629-5G
COBBAM-1629-1GEA
0
EUR
COBBAM-1629-1G
COBBAM-1629-25G
COBBAM-1629-5G
Methyltetrahydropyran-4-ylacetat
Methyltetrahydropyran-4-ylacetat
Formel:
C₈H₁₄O₃ |
MDL:
MFCD09972087 CAS-Nummer: 156002-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...