1-(4-Bromphenyl)cyclopropanamin

Lieferant: COMBI-BLOCKS

AM-1631-1G
COBBAM-1631-1GEA 359 EUR
COBBAM-1631-1G
1-(4-Bromphenyl)cyclopropanamin
1-(4-Bromphenyl)cyclopropanamin
Formel: C₉H₁₀BrN
Molecular Weight: 212,0900 g/mol
MDL: MFCD07374473
CAS-Nummer: 345965-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR