1-Boc-piperidin-3-carbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
N-Boc-DL-nipecotic acid, 1-[(tert-butoxy)carbonyl]piperidine-3-carboxylic acid, 1-Boc-Nipecotic acid, Boc-Nip-OH, 1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid, N-BOC-3-Piperidinecarboxylic acid
AM-1708-5G
AM-1708-25G
AM-1708-100G
COBBAM-1708-5GEA
0
EUR
COBBAM-1708-5G
COBBAM-1708-25G
COBBAM-1708-100G
1-Boc-piperidin-3-carbonsäure
1-Boc-piperidin-3-carbonsäure
Formel:
C₁₁H₁₉NO₄ |
MDL:
MFCD00673775 CAS-Nummer: 84358-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...