(S)-(+)-1-Boc-3-hydroxypyrrolidin

Lieferant: COMBI-BLOCKS

Synonyme: tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate, (S)-tert-Butyl 3-hydroxypyrrolidine-1-carboxylate

AM-1756-5G AM-1756-25G
COBBAM-1756-5GEA 0 EUR
COBBAM-1756-5G COBBAM-1756-25G
(S)-(+)-1-Boc-3-hydroxypyrrolidin
(S)-(+)-1-Boc-3-hydroxypyrrolidin
Formel: C₉H₁₇NO₃
MDL: MFCD01317839
CAS-Nummer: 101469-92-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR