1-Boc-4-(methylamino)piperidin

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Methylamino)piperidine, N1-BOC protected, tert-Butyl 4-(methylamino)piperidine-1-carboxylate

AM-1762-5G AM-1762-25G AM-1762-1G
COBBAM-1762-5GEA 0 EUR
COBBAM-1762-5G COBBAM-1762-25G COBBAM-1762-1G
1-Boc-4-(methylamino)piperidin
1-Boc-4-(methylamino)piperidin
Formel: C₁₁H₂₂N₂O₂
MDL: MFCD02259411
CAS-Nummer: 147539-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR