(S)-(-)-1-Boc-3-aminopyrrolidin
Lieferant: COMBI-BLOCKS
Synonyme:
(S)-1-Boc-3-Aminopyrrolidine, (S)-(-)-1-BOC-3-aminopyrrolidine, tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate
AM-1768-1G
AM-1768-5G
COBBAM-1768-1GEA
0
EUR
COBBAM-1768-1G
COBBAM-1768-5G
(S)-(-)-1-Boc-3-aminopyrrolidin
(S)-(-)-1-Boc-3-aminopyrrolidin
Formel:
C₉H₁₈N₂O₂ |
MDL:
MFCD03419271 CAS-Nummer: 147081-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...