1-(3-Bromphenyl)-2-piperidinon

Lieferant: COMBI-BLOCKS

AM-2085-5G
COBBAM-2085-5GEA 797 EUR
COBBAM-2085-5G
1-(3-Bromphenyl)-2-piperidinon
1-(3-Bromphenyl)-2-piperidinon
Formel: C₁₁H₁₂BrNO
Molecular Weight: 254,123 g/mol
MDL: MFCD04037063
CAS-Nummer: 1016847-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR