2-Ethoxy-4-fluor-1-nitrobenzol
Lieferant: COMBI-BLOCKS
AN-1019-5G
AN-1019-1G
AN-1019-25G
COBBAN-1019-5GEA
352
EUR
COBBAN-1019-5G
COBBAN-1019-1G
COBBAN-1019-25G
2-Ethoxy-4-fluor-1-nitrobenzol
2-Ethoxy-4-fluor-1-nitrobenzol
Formel:
C₈H₈FNO₃ Molecular Weight: 185.1524 g/mol |
MDL:
MFCD04115630 CAS-Nummer: 28987-44-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...