3-Fluor-4-nitroanisol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-4-methoxy-1-nitrobenzene

AN-1026-5G AN-1026-25G AN-1026-1G
COBBAN-1026-5GEA 0 EUR
COBBAN-1026-5G COBBAN-1026-25G COBBAN-1026-1G
3-Fluor-4-nitroanisol
3-Fluor-4-nitroanisol
Formel: C₇H₆FNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04115632
CAS-Nummer: 446-38-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR