4-Fluor-3-nitrobenzonitril

Lieferant: COMBI-BLOCKS

AN-1043-5G AN-1043-25G AN-1043-1G
COBBAN-1043-5GEA 46 EUR
COBBAN-1043-5G COBBAN-1043-25G COBBAN-1043-1G
4-Fluor-3-nitrobenzonitril
4-Fluor-3-nitrobenzonitril
Formel: C₇H₃FN₂O₂
Molecular Weight: 166.11 g/mol
MDL: MFCD01632197
CAS-Nummer: 1009-35-4
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR