4-Brom-3-chlorphenol

Lieferant: COMBI-BLOCKS

AN-1044-1G AN-1044-5G AN-1044-25G
COBBAN-1044-1GEA 0 EUR
COBBAN-1044-1G COBBAN-1044-5G COBBAN-1044-25G
4-Brom-3-chlorphenol
4-Brom-3-chlorphenol
Formel: C₆H₄BrClO
Molecular Weight: 207.45 g/mol
MDL: MFCD00070744
CAS-Nummer: 13631-21-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR