2-Chlor-5-nitrobenzonitril
Lieferant: COMBI-BLOCKS
AN-1050-5G
AN-1050-100G
AN-1050-25G
COBBAN-1050-5GEA
0
EUR
COBBAN-1050-5G
COBBAN-1050-100G
COBBAN-1050-25G
2-Chlor-5-nitrobenzonitril
2-Chlor-5-nitrobenzonitril
Formel:
C₇H₃ClN₂O₂ Siedepunkt: 119…122 °C (0,6 mmHg) Schmelzpunkt: 108 °C |
MDL:
MFCD00007292 CAS-Nummer: 16588-02-6 EINECS: 240-643-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...