1-Brom-4-chlor-2-nitrobenzol
Lieferant: COMBI-BLOCKS
|
Achtung
|
AN-1055-25G
AN-1055-5G
AN-1055-100G
COBBAN-1055-25GEA
0
EUR
COBBAN-1055-25G
COBBAN-1055-5G
COBBAN-1055-100G
1-Brom-4-chlor-2-nitrobenzol
1-Brom-4-chlor-2-nitrobenzol
Formel:
C₆H₃BrClNO₂ Schmelzpunkt: 70 °C Dichte: 2,048 g/cm³ (20 °C) |
MDL:
MFCD00024320 CAS-Nummer: 41513-04-6 EINECS: 255-421-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...