4-Cyan-2-fluoranilin

Lieferant: COMBI-BLOCKS

AN-1059-100G AN-1059-25G AN-1059-5G
COBBAN-1059-100GEA 0 EUR
COBBAN-1059-100G COBBAN-1059-25G COBBAN-1059-5G
4-Cyan-2-fluoranilin
4-Cyan-2-fluoranilin
Formel: C₇H₅FN₂
MDL: MFCD00055559
CAS-Nummer: 63069-50-1
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR