1,4-Dibrom-2-nitrobenzol
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
2,5-Dibromonitrobenzene
AN-1087-5G
AN-1087-25G
COBBAN-1087-5GEA
0
EUR
COBBAN-1087-5G
COBBAN-1087-25G
1,4-Dibrom-2-nitrobenzol
1,4-Dibrom-2-nitrobenzol
Formel:
C₆H₃Br₂NO₂ Molecular Weight: 280,9 g/mol Schmelzpunkt: 81…85 °C Dichte: 2,374 g/cm³ (20 °C) |
MDL:
MFCD00007046 CAS-Nummer: 3460-18-2 EINECS: 222-404-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...