3-Fluor-p-anisidin

Lieferant: COMBI-BLOCKS

AN-1094-5G AN-1094-25G
COBBAN-1094-5GEA 31 EUR
COBBAN-1094-5G COBBAN-1094-25G
3-Fluor-p-anisidin
3-Fluor-p-anisidin
Formel: C₇H₈FNO
MDL: MFCD00075040
CAS-Nummer: 366-99-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR