2-Brom-6-fluor-4-nitroanisol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-5-fluoro-4-methoxynitrobenzene

AN-1095-5G
COBBAN-1095-5GEA 124 EUR
COBBAN-1095-5G
2-Brom-6-fluor-4-nitroanisol
2-Brom-6-fluor-4-nitroanisol
Formel: C₇H₅BrFNO₃
Molecular Weight: 250,02 g/mol
MDL: MFCD08062383
CAS-Nummer: 875664-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR