1-Cyclohexyl-4-nitrobenzol

Lieferant: COMBI-BLOCKS

AN-1098-5G AN-1098-1G
COBBAN-1098-5GEA 595 EUR
COBBAN-1098-5G COBBAN-1098-1G
1-Cyclohexyl-4-nitrobenzol
1-Cyclohexyl-4-nitrobenzene
Formel: C₁₂H₁₅NO₂
MDL: MFCD00130008
CAS-Nummer: 5458-48-0
EINECS: 226-720-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR