2-Chlor-3-fluoranilin
Lieferant: COMBI-BLOCKS
AN-1124-1G
AN-1124-25G
AN-1124-5G
COBBAN-1124-1GEA
0
EUR
COBBAN-1124-1G
COBBAN-1124-25G
COBBAN-1124-5G
2-Chlor-3-fluoranilin
2-Chlor-3-fluoranilin
Formel:
C₆H₅ClFN |
MDL:
MFCD03407961 CAS-Nummer: 21397-08-0 EINECS: 244-366-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...