2-Chlor-3-fluoranilin

Lieferant: COMBI-BLOCKS

AN-1124-1G AN-1124-25G AN-1124-5G
COBBAN-1124-1GEA 0 EUR
COBBAN-1124-1G COBBAN-1124-25G COBBAN-1124-5G
2-Chlor-3-fluoranilin
2-Chlor-3-fluoranilin
Formel: C₆H₅ClFN
MDL: MFCD03407961
CAS-Nummer: 21397-08-0
EINECS: 244-366-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR