3-Hydroxy-5-nitrobenzonitril

Lieferant: COMBI-BLOCKS

AN-1132-1G AN-1132-5G
COBBAN-1132-1GEA 0 EUR
COBBAN-1132-1G COBBAN-1132-5G
3-Hydroxy-5-nitrobenzonitril
3-Hydroxy-5-nitrobenzonitril
Formel: C₇H₄N₂O₃
MDL: MFCD09258662
CAS-Nummer: 929000-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR