(N,N-Bis-Boc)-5-brom-2-chloranilin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2-chloroaniline, N,N-Bis-BOC protected

AN-1133-1G
COBBAN-1133-1GEA 656 EUR
COBBAN-1133-1G
(N,N-Bis-Boc)-5-brom-2-chloranilin
(N,N-Bis-Boc)-5-brom-2-chloranilin
Formel: C₁₆H₂₁BrClNO₄
MDL: MFCD09258633
CAS-Nummer: 929000-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR