1,3-Dibrom-5-nitrobenzol

Lieferant: COMBI-BLOCKS

AN-1143-5G AN-1143-1G
COBBAN-1143-5GEA 232 EUR
COBBAN-1143-5G COBBAN-1143-1G
1,3-Dibrom-5-nitrobenzol
1,3-Dibromo-5-nitrobenzene
Formel: C₆H₃Br₂NO₂
Molecular Weight: 280.9 g/mol
MDL: MFCD00092529
CAS-Nummer: 6311-60-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR