5-Methyl-2-nitroanisol
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
1-Methoxy-3-methyl-6-nitrobenzol, 3-Methoxy-4-nitrotoluol, Methyl-6-nitro-m-tolylether
AN-1146-5G
AN-1146-100G
AN-1146-25G
COBBAN-1146-5GEA
0
EUR
COBBAN-1146-5G
COBBAN-1146-100G
COBBAN-1146-25G
5-Methyl-2-nitroanisol
5-Methyl-2-nitroanisol
Formel:
C₈H₉NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007367 CAS-Nummer: 38512-82-2 EINECS: 253-975-1 UN: 2446 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...