2-(Methylthio)-5-nitrobenzonitril

Lieferant: COMBI-BLOCKS

AN-1150-5G AN-1150-1G
COBBAN-1150-5GEA 303 EUR
COBBAN-1150-5G COBBAN-1150-1G
2-(Methylthio)-5-nitrobenzonitril
2-(Methylthio)-5-nitrobenzonitril
Formel: C₈H₆N₂O₂S
MDL: MFCD09475845
CAS-Nummer: 74631-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR