2,3-Difluor-4-nitroanisol
Lieferant: COMBI-BLOCKS
AN-1156-5G
AN-1156-25G
AN-1156-1G
COBBAN-1156-5GEA
76.4
EUR
COBBAN-1156-5G
COBBAN-1156-25G
COBBAN-1156-1G
2,3-Difluor-4-nitroanisol
2,3-Difluor-4-nitroanisol
Formel:
C₇H₅F₂NO₃ Molecular Weight: 189.12 g/mol |
MDL:
MFCD07995055 CAS-Nummer: 66684-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...