2-Chlor-6-nitroanisol

Lieferant: COMBI-BLOCKS

AN-1165-5G AN-1165-1G
COBBAN-1165-5GEA 102 EUR
COBBAN-1165-5G COBBAN-1165-1G
2-Chlor-6-nitroanisol
2-Chlor-6-nitroanisol
Formel: C₇H₆ClNO₃
Molecular Weight: 187.58 g/mol
MDL: MFCD00065066
CAS-Nummer: 80866-77-9
EINECS: 279-580-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR