4-Chlor-2-(methoxymethyl)-N-methylanilin

Lieferant: COMBI-BLOCKS

AN-1169-1G
COBBAN-1169-1GEA 1980 EUR
COBBAN-1169-1G
4-Chlor-2-(methoxymethyl)-N-methylanilin
4-Chlor-2-(methoxymethyl)-N-methylanilin
Formel: C₉H₁₂ClNO
MDL: MFCD09475903
CAS-Nummer: 941294-12-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR