4-Fluor-2-methoxy-N-methylanilin

Lieferant: COMBI-BLOCKS

AN-1171-1G
COBBAN-1171-1GEA 208 EUR
COBBAN-1171-1G
4-Fluor-2-methoxy-N-methylanilin
4-Fluor-2-methoxy-N-methylanilin
Formel: C₈H₁₀FNO
MDL: MFCD09475904
CAS-Nummer: 941294-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR