4-(4-Fluor-2-nitrophenyl)morpholin

Lieferant: COMBI-BLOCKS

AN-1202-5G AN-1202-25G AN-1202-1G
COBBAN-1202-5GEA 166 EUR
COBBAN-1202-5G COBBAN-1202-25G COBBAN-1202-1G
4-(4-Fluor-2-nitrophenyl)morpholin
4-(4-Fluor-2-nitrophenyl)morpholin
Formel: C₁₀H₁₁FN₂O₃
Molecular Weight: 226.20 g/mol
MDL: MFCD00798369
CAS-Nummer: 238418-75-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR