1-(4-Fluor-2-nitrophenyl)piperidin
Lieferant: COMBI-BLOCKS
AN-1225-5G
AN-1225-25G
AN-1225-1G
COBBAN-1225-5GEA
127
EUR
COBBAN-1225-5G
COBBAN-1225-25G
COBBAN-1225-1G
1-(4-Fluor-2-nitrophenyl)piperidin
1-(4-Fluor-2-nitrophenyl)piperidin
Formel:
C₁₁H₁₃FN₂O₂ Molecular Weight: 224.23 g/mol |
MDL:
MFCD00798370 CAS-Nummer: 719-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...