N-Cyclopropyl-4-fluor-2-nitroanilin

Lieferant: COMBI-BLOCKS

AN-1226-5G
COBBAN-1226-5GEA 141 EUR
COBBAN-1226-5G
N-Cyclopropyl-4-fluor-2-nitroanilin
N-Cyclopropyl-4-fluor-2-nitroanilin
Formel: C₉H₉FN₂O₂
Molecular Weight: 196,18 g/mol
MDL: MFCD08448315
CAS-Nummer: 887351-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR