3-Amino-4-methylbenzotrifluorid

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methyl-5-(trifluoromethyl)aniline

AN-1247-25G
COBBAN-1247-25GEA 303 EUR
COBBAN-1247-25G
3-Amino-4-methylbenzotrifluorid
3-Amino-4-methylbenzotrifluorid
Formel: C₈H₈F₃N
Molecular Weight: 175,16 g/mol
MDL: MFCD00134580
CAS-Nummer: 25449-96-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR