2'-Fluor-4'-iodacetanilid

Lieferant: COMBI-BLOCKS

AN-1249-5G AN-1249-1G
COBBAN-1249-5GEA 111 EUR
COBBAN-1249-5G COBBAN-1249-1G
2'-Fluor-4'-iodacetanilid
2'-Fluor-4'-iodacetanilid
Formel: C₈H₇FINO
MDL: MFCD04039298
CAS-Nummer: 97760-94-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR