3',5'-Dibromacetanilid

Lieferant: COMBI-BLOCKS

AN-1270-5G AN-1270-1G
COBBAN-1270-5GEA 0 EUR
COBBAN-1270-5G COBBAN-1270-1G
3',5'-Dibromacetanilid
3',5'-Dibromacetanilid
Formel: C₈H₇Br₂NO
MDL: MFCD09878317
CAS-Nummer: 119430-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR