4'-Brom-3'-chlor-2'-methylacetanilid

Lieferant: COMBI-BLOCKS

AN-1278-5G AN-1278-1G
COBBAN-1278-5GEA 0 EUR
COBBAN-1278-5G COBBAN-1278-1G
4'-Brom-3'-chlor-2'-methylacetanilid
4'-Brom-3'-chlor-2'-methylacetanilid
Formel: C₉H₉BrClNO
MDL: MFCD08059123
CAS-Nummer: 125328-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR