4-Brom-2-nitrophenol

Lieferant: COMBI-BLOCKS

AN-1280-25G AN-1280-5G AN-1280-100G
COBBAN-1280-25GEA 122 EUR
COBBAN-1280-25G COBBAN-1280-5G COBBAN-1280-100G
4-Brom-2-nitrophenol
4-Brom-2-nitrophenol
Formel: C₆H₄BrNO₃
Molecular Weight: 218.01 g/mol
Siedepunkt: 259
Schmelzpunkt: 90…94 °C
MDL: MFCD00082540
CAS-Nummer: 7693-52-9
EINECS: 231-707-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR