4-Brom-2-nitrophenol
Lieferant: COMBI-BLOCKS
AN-1280-25G
AN-1280-5G
AN-1280-100G
COBBAN-1280-25GEA
122
EUR
COBBAN-1280-25G
COBBAN-1280-5G
COBBAN-1280-100G
4-Brom-2-nitrophenol
4-Brom-2-nitrophenol
Formel:
C₆H₄BrNO₃ Molecular Weight: 218.01 g/mol Siedepunkt: 259 Schmelzpunkt: 90…94 °C |
MDL:
MFCD00082540 CAS-Nummer: 7693-52-9 EINECS: 231-707-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...