4-Nitro-3-(trifluormethyl)anisol

Lieferant: COMBI-BLOCKS

AN-1285-5G AN-1285-25G AN-1285-1G
COBBAN-1285-5GEA 0 EUR
COBBAN-1285-5G COBBAN-1285-25G COBBAN-1285-1G
4-Nitro-3-(trifluormethyl)anisol
4-Nitro-3-(trifluormethyl)anisol
Formel: C₈H₆F₃NO₃
MDL: MFCD00053611
CAS-Nummer: 344-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR