1,2-Dibrom-4-nitrobenzol

Lieferant: COMBI-BLOCKS

AN-1296-25G AN-1296-1G AN-1296-5G
COBBAN-1296-25GEA 0 EUR
COBBAN-1296-25G COBBAN-1296-1G COBBAN-1296-5G
1,2-Dibrom-4-nitrobenzol
1,2-Dibromo-4-nitrobenzene
Formel: C₆H₃Br₂NO₂
MDL: MFCD00055373
CAS-Nummer: 5411-50-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR