4-Nitro-2-(trifluormethyl)anisol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxy-5-nitrobenzotrifluorid, 1-Methoxy-4-nitro-2-(trifluormethyl)benzol

AN-1319-25G AN-1319-100G
COBBAN-1319-25GEA 61.9 EUR
COBBAN-1319-25G COBBAN-1319-100G
4-Nitro-2-(trifluormethyl)anisol
4-Nitro-2-(trifluormethyl)anisol
Formel: C₈H₆F₃NO₃
Molecular Weight: 221.14 g/mol
Schmelzpunkt: 79…79.5 °C
MDL: MFCD00007328
CAS-Nummer: 654-76-2
EINECS: 211-507-3
UN: 2811
ADR: 6.1 I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR