1-Brom-3,5-dinitrobenzol

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dinitrobrombenzol

AN-1333-5G AN-1333-1G
COBBAN-1333-5GEA 169 EUR
COBBAN-1333-5G COBBAN-1333-1G
1-Brom-3,5-dinitrobenzol
1-Bromo-3,5-dinitrobenzene
Formel: C₆H₃BrN₂O₄
Molecular Weight: 247 g/mol
MDL: MFCD00156596
CAS-Nummer: 18242-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR