2,3-Difluor-6-nitroanisol

Lieferant: COMBI-BLOCKS

Synonyme: 1,2-Difluoro-3-methoxy-4-nitrobenzene

AN-1347-5G AN-1347-25G AN-1347-100G
COBBAN-1347-5GEA 38.5 EUR
COBBAN-1347-5G COBBAN-1347-25G COBBAN-1347-100G
2,3-Difluor-6-nitroanisol
2,3-Difluor-6-nitroanisol
Formel: C₇H₅F₂NO₃
Molecular Weight: 189.1163 g/mol
MDL: MFCD03840468
CAS-Nummer: 66684-60-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR